Learn More
SV40 Large T Antigen Ab-1 Mouse Monoclonal Antibody, Epredia™
Epredia™ SV40 Large T Antigen Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.
Marke: Epredia MS-1832-P0
Beschreibung
The simian virus 40 large T antigen plays important role in SV40 transfection and transformation of the cells. It binds to Rb and p53 suppressor proteins leading to the inactivation of suppressor pathway.Host Species: Mouse
Clone: PAb101; originally clone 412
Isotype: IgG2a1
Species Reactivity: SV40 virus.
Epitope: C-terminal
Immunogen: SV40-transformed mouse line B4
Molecular Weight: 94kDa
Positive Control: SV40-transfected cells
Recommended for:
- Western Blotting
- Immunofluorescence
- Immunoprecipitation
Note:
USA: RUO; Int'l: RUO
Spezifikation
| SV40 Large T Antigen Ab-1 | |
| Monoclonal | |
| 200μg/mL | |
| Mouse | |
| 100 μL | |
| Infectious Disease | |
| Simian virus 40 (SV40) |
| Immunofluorescence, Immunoprecipitation, Western Blot | |
| PAb101; same as clone 412 | |
| Unconjugated | |
| SV40 transformed mouse line B4 | |
| RUO | |
| Primary | |
| Concentrated |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.