Learn More
TAG-72/CA 72-4TM Ab-1 Mouse Monoclonal Antibody, Epredia™
Provide accurate, reproducible results with the Epredia™ TAG-72/CA 72-4 Ab-1, Mouse Monoclonal Antibody.
Marke: Epredia MS-138-PABX
Weitere Details : Netto-Gewicht : 1.00000kg
Beschreibung
Tumor-associated glycoprotein (TAG-72) is an oncofetal antigen with properties of a mucin. About 60% of carcinoma patients express TAG-72 in their sera.
Host Species: Mouse
Clone: B72.3
Isotype: IgG1/κ
Species Reactivity: Human, Cow, Dog, Hamster and Rat. Others not known.
Epitope: Mucin-carried-sialylated-Tn epitope
Immunogen: Membrane-enriched fraction of a human breast carcinoma liver metastasis
Molecular Weight: >1000kDa
Positive Control: Breast or lung CA
Cellular Localization: Cell surface and cytoplasmic. The majority of human adenocarcinomas including colorectal, pancreatic, gastric, ovarian, endometrial, mammary, and non-small cell lung cancer display some cell populations that are positive for Ab-1 staining. Weak or no reactivity has been observed with most cell types of normal adult tissue excepting the secretory endometrium. Ab-1 is reportedly useful in distinguishing pulmonary adenocarcinomas (B72.3+) from pleural mesotheliomas (B72.3-).
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Flow Cytometry
- Radioimaging
Note:
USA: RUO; Int'l: RUO
Spezifikation
TAG-72/CA 72-4 Ab-1 | |
Monoclonal | |
1mg/mL | |
Mouse | |
200 μL | |
Cancer and Tumor Biology | |
Bovine, Canine, Hamster, Human, Rat | |
IgG2a κ |
Flow Cytometry, Immunohistochemistry (Paraffin) | |
B72.3 | |
Unconjugated | |
Membrane-enriched fraction of a human breast carcinoma liver metastasis | |
RUO | |
Primary | |
Concentrated |