Learn More
Epredia™ Lab Vision™ Tartrate Resistant Acid Phosphatase (TRAP) Ab-1, Mouse Monoclonal Antibody
Tartrate Resistant Acid Phosphatase (TRAP) Ab-1, Mouse MAb is one marker in a comprehensive ab portfolio that addresses all major disease states, ensuring the ideal tests are always available.
Marke: Epredia™ MS-1768-S1
Beschreibung
Tartrate-resistant acid phosphatase (TRAP) is a basic iron-binding protein with high activity towards phosphoproteins, ATP and 4-nitrophenyl phosphatase. It is detected in human alveolar macrophages, osteoclasts, spleen and liver. Its expression is increased in spleen and monocytes of patients with Gaucher's disease; spleenocytes and circulating leukocytes of patients with hairy cell leukemia; spleen of patients with Hodgkin's lymphoma and in the sera of patients undergoing active bone turnover.Host Species: Mouse
Clone: 26E5
Isotype: IgG2b
Species Reactivity: Human. Others not known.
Epitope: N-terminal
Immunogen: Recombinant protein corresponding to N-terminal region of human TRAP
Positive Control: Lung (macrophages)
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Spezifikation
| Tartrate Resistant Acid Phosphatase (TRAP) Ab-1 | |
| Monoclonal | |
| Unconjugated | |
| Recombinant protein corresponding to N-terminal region of human TRAP | |
| 500 μL | |
| Cancer and Tumor Biology | |
| Human | |
| IgG2b |
| Immunohistochemistry (Paraffin) | |
| 26E5 | |
| Mouse | |
| Lung (macrophages) | |
| IVD | |
| Primary | |
| Concentrated |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.