Learn More
Epredia™ Lab Vision™ Tartrate Resistant Acid Phosphatase (TRAP) Ab-1, Mouse Monoclonal Antibody
Tartrate Resistant Acid Phosphatase (TRAP) Ab-1, Mouse MAb is one marker in a comprehensive ab portfolio that addresses all major disease states, ensuring the ideal tests are always available.
110.16 € - 520.20 €
Spezifikation
| Antigen | Tartrate Resistant Acid Phosphatase (TRAP) Ab-1 |
|---|---|
| Klon | 26E5 |
| Anwendungen | Immunohistochemistry (Paraffin) |
| Klassifikation | Monoclonal |
| Konjugat | Unconjugated |
| Produktcode | Marke | Menge | Form | Preis | Menge & Verfügbarkeit | ||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Form | Preis | Menge & Verfügbarkeit | ||||
|
12630147
|
Epredia™
MS-1768-S0 |
100 μL | Concentrated | ||||||
|
12640147
|
Epredia™
MS-1768-S1 |
500 μL | Concentrated |
473.00 €
500 Mikroliter |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
|
12610147
|
Epredia™
MS-1768-S |
1 mL | Concentrated | ||||||
|
12620147
|
Epredia™
MS-1768-R7 |
7 mL | Prediluted | ||||||
Beschreibung
Tartrate-resistant acid phosphatase (TRAP) is a basic iron-binding protein with high activity towards phosphoproteins, ATP and 4-nitrophenyl phosphatase. It is detected in human alveolar macrophages, osteoclasts, spleen and liver. Its expression is increased in spleen and monocytes of patients with Gaucher's disease; spleenocytes and circulating leukocytes of patients with hairy cell leukemia; spleen of patients with Hodgkin's lymphoma and in the sera of patients undergoing active bone turnover.Host Species: Mouse
Clone: 26E5
Isotype: IgG2b
Species Reactivity: Human. Others not known.
Epitope: N-terminal
Immunogen: Recombinant protein corresponding to N-terminal region of human TRAP
Positive Control: Lung (macrophages)
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Spezifikation
| Tartrate Resistant Acid Phosphatase (TRAP) Ab-1 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| IVD | |
| Human | |
| IgG2b |
| 26E5 | |
| Monoclonal | |
| Mouse | |
| Cancer and Tumor Biology | |
| Recombinant protein corresponding to N-terminal region of human TRAP | |
| Primary |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.