missing translation for 'onlineSavingsMsg'
Learn More

TTF-1 (Thyroid Transcription Factor-1) Ab-1 Mouse Monoclonal Antibody, Epredia™

Provide accurate, reproducible results with the Epredia™ TTF-1 (Thyroid Transcription Factor-1) Ab-1, Mouse Monoclonal Antibody.

Marke:  Epredia MS-699-P1

Weitere Details : Netto-Gewicht : 0.00190kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13221223

  • 442.00 € / 500 Mikroliter
Verfügbar ab: 09-07-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

TTF-1(Thyroid transcription factor-1) is a member of the NKx2 family of homeodomain transcription factors. It is expressed in epithelial cells of the thyroid gland and the lung. Nuclei from liver, stomach, pancreas, small intestine, colon, kidney, breast, skin, testes, pituitary, prostate, and adrenal glands are unreactive. TTF-1 is detected in primary lung adenocarcinomas and small cell carcinomas and is absent in colon and breast carcinomas. Staining with TTF-1 antibody is useful for distinguishing between tumors of lung and non-lung origin.

Host Species: Mouse

Clone: 8G7G3/1

Isotype: IgG1/κ

Species Reactivity: Human, Mouse and Rat. Shows a broad species reactivity.

Epitope: Not determined

Immunogen: Rat TTF-1 recombinant protein

Molecular Weight: 40kDa

Positive Control: MAD109 cells, normal thyroid or lung

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunofluorescence
Spezifikation

Spezifikation

TTF-1 (Thyroid Transcription Factor-1) Ab-1
Monoclonal
200μg/mL
Mouse
500 μL
Transcription Regulation
Human, Mouse, Rat
IgG1 κ
Immunofluorescence, Immunohistochemistry (Paraffin)
8G7G3/1
Unconjugated
Rat TTF-1 recombinant protein
IVD
Primary
Purified
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote