Learn More
LACTB Mouse anti-Human, CoraLite 488, Clone: 3C3C3, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech CL488-66785-100UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
LACTB is also named as MRPL56,whic is a component of the large subunit of the mitochondrial ribosome that is encoded by the nuclear genome.LACTB is a mammalian active-site serine protein that has evolved from a bacterial penicillin-binding protein.It promotes intramitochondrial membrane organization and micro-compartmentalization through polymerization(PMID:19858488). Northern blot analysis of mouse tissues detected expression at high levels in liver, heart, and skeletal muscle, with lower levels in kidney, testis, brain, lung, and spleen(PMID:11707067). The full-length of LACTB is about 61kDa, and a 115aa transit peptide can be removed in the mature form.
Spezifikation
LACTB | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Immunofluorescence | |
3C3C3 | |
CoraLite 488 | |
100 μL | |
114294 | |
Liquid |