Learn More
LARP2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA540915
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: RNTRTPRTPR TPQLKDSSQT SRFYPVVKEG RTLDAKMPRK RKTRHSSNPP Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 100%; Rat: 100%; Zebrafish: 79%
LARP2 Polyclonal antibody specifically detects LARP2 in Human samples. It is validated for Immunohistochemistry, Western Blot.Spezifikation
LARP2 | |
Polyclonal | |
Unconjugated | |
LARP1B | |
1700108L22Rik; 4933421B21Rik; AA690246; DKFZp686L13217; fc76b04; la related protein; La ribonucleoprotein 1B; La ribonucleoprotein domain family member 1B; La ribonucleoprotein domain family member 2; La ribonucleoprotein domain family, member 1B; La ribonucleoprotein domain family, member 2; La-like protein 1B-like protein; la-related protein 1; la-related protein 1B; la-related protein 1B; La-related protein 2; LARP; LARP1B; LARP2; MGC117277; MGC75174; RGD1307509; wu:fc76b04 | |
Rabbit | |
Protein A | |
RUO | |
55132 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q659C4 | |
LARP1B | |
Synthetic peptide directed towards the middle region of human LARP2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |