Learn More
LARS Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA559338
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: KWDTERVFEV NASNLEKQTS KGKYFVTFPY PYMNGRLHLG HTFSLSKCEF AVGYQRLKGK CCLFPFGLHC TGMPIKACAD KLK Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 89%.
This gene encodes a cytosolic leucine-tRNA synthetase, a member of the class I aminoacyl-tRNA synthetase family. The encoded enzyme catalyzes the ATP-dependent ligation of L-leucine to tRNA(Leu). It is found in the cytoplasm as part of a multisynthetase complex and interacts with the arginine tRNA synthetase through its C-terminal domain. Alternatively spliced transcript variants of this gene have been found; however, their full-length nature is not known.Spezifikation
LARS | |
Polyclonal | |
Unconjugated | |
LARS | |
HSPC192, LARS1, LEURS, LEUS, LRS, PIG44, RNTLS, hr025Cl, cytoplasmic leucyl-tRNA synthetase, cytosolic leucyl-tRNA synthetase, leucine tRNA ligase 1, cytoplasmic, leucine translase, leucine--tRNA ligase, cytoplasmic, leucine-tRNA ligase, leucyl-tRNA synthetase, cytoplasmic, proliferation-inducing gene 44 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51520 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9P2J5 | |
LARS | |
Recombit protein corresponding to Human LARS | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only