missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lass4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA550695
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
The antibody detects endogenous levels of total Lass4 protein.
Lass4 Polyclonal antibody specifically detects Lass4 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
Lass4 | |
Polyclonal | |
Unconjugated | |
CERS4 | |
2900019C14Rik; Ceramide synthase 4; CerS4; FLJ12089; LAG1 homolog, ceramide synthase 4; LAG1 longevity assurance homolog 4; Lass4; longevity assurance homolog 4; Sphingosine N-acyltransferase CERS4; TRAM homolog 1; translocating chain-associating membrane protein homolog 1; Trh1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79603 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
2.3 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q9HA82 | |
CERS4 | |
Synthetic peptide corresponding to a region derived from internal residues of human ceramide synthase 4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |