Learn More
LENG1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA560412
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: PDEKIKSRLD PLREMQKHLG KKRQHGGDEG SRSRKEKEGS EKQRPKEPPS LDQLRAERLR REAA Highest antigen sequence identity to the following orthologs: Mouse - 66%, Rat - 69%.
The function of this protein remains unknown.Spezifikation
LENG1 | |
Polyclonal | |
Unconjugated | |
LENG1 | |
1500001K17Rik; Leng1; leukocyte receptor cluster (LRC) member 1; leukocyte receptor cluster member 1; Leukocyte receptor cluster member 1 homolog | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79165 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96BZ8 | |
LENG1 | |
Recombinant protein corresponding to Human LENG1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only