missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Lithiumdodecylsulfat, 98 %

Click to view available options
Menge:
5g
25g
100g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
2044-56-6
C12H25LiO4S
272.33
MFCD00007467
YFVGRULMIQXYNE-UHFFFAOYSA-M
lithium dodecyl sulfate, lithium dodecylsulfate, sulfuric acid, monododecyl ester, lithium salt, lauryl sulfate lithium salt, lithium lauryl sulfate, dodecyl lithium sulfate, lds, lithium 1+ ion dodecyl sulfate, sulfuric acid, monododecyl ester, lithium salt 1:1, lithium dodecyl sulfate, ultra pure
2735071
Lithium(1+) Dodecylsulfat
[Li+].CCCCCCCCCCCCOS([O-])(=O)=O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
2044-56-6
C12H25LiO4S
272.33
MFCD00007467
YFVGRULMIQXYNE-UHFFFAOYSA-M
lithium dodecyl sulfate, lithium dodecylsulfate, sulfuric acid, monododecyl ester, lithium salt, lauryl sulfate lithium salt, lithium lauryl sulfate, dodecyl lithium sulfate, lds, lithium 1+ ion dodecyl sulfate, sulfuric acid, monododecyl ester, lithium salt 1:1, lithium dodecyl sulfate, ultra pure
2735071
Lithium(1+) Dodecylsulfat
[Li+].CCCCCCCCCCCCOS([O-])(=O)=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemischer Name oder Material Lithium dodecylsulfate
Menge 5g
UN-Nummer UN1325
Formelmasse 272.33
Reinheit (%) 98%
Geruch Odorless

RUO – Research Use Only