Learn More
LMO2 Recombinant Rabbit Monoclonal Antibody (13H1L2), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Marke: Invitrogen 701156
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody is predicted to react with non-human primate, mouse, rabbit and rat based on sequence homology. Intact IgG appears on a non-reducing gel as ∽150 kDa band and upon reduction generating a ∽25 kDa light chain band and a ∽50 kDa heavy chain. Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
LMO2 Recombinant Monoclonal antibody specifically detects LMO2 in Human samples. It is validated for Western Blot.Spezifikation
LMO2 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
cysteine-rich protein TTG-2; LIM domain only 2; LIM domain only 2 (rhombotin-like 1); LIM domain only protein 2; LIM domain only-2; LIM only 2; LMO2; LMO-2; LMO2 protein; Rbtn2; Rbtn-2; RBTNL1; RHOM2; Rhom-2; rhombotin 2; rhombotin-2; rhombotin-like 1; si:dkey-10o6.5; T-cell translocation gene 2; T-cell translocation protein 2; TTG2; wu:fc83a08; zgc:111930; zlmo2 | |
Recombinant protein corresponding to amino acids 1–127 of human LMO2 | |
100 μg | |
Primary | |
Human | |
Liquid |
Western Blot | |
13H1L2 | |
Unconjugated | |
P25791 | |
Rabbit | |
Protein A | |
RUO | |
4005 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG |