Learn More
LMOD3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 14948-1-AP-20UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
This antibody was raised against the N-terminal region of human LMOD3. It detects a double bands around 80 kDa and 65 kDa in heart and skeletal muscle lysates. The reason causing the discrepancy between the predicted and observed molecular weight is not c
The function of this protein remains unknown.Spezifikation
LMOD3 | |
Polyclonal | |
Unconjugated | |
LMOD3 | |
Leiomodin 3, leiomodin 3 (fetal), Leiomodin, fetal form, LMOD3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
320502, 500267, 56203 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
E9QA62, Q0VAK6 | |
Lmod3 | |
LMOD3 Fusion Protein Ag6758 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |