Learn More
LYPLA1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA528034
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: LYPLA1 tansfected 293T cell. Predicted reactivity: Mouse (91%), Rat (92%), Xenopus laevis (80%), Rabbit (97%), Sheep (94%), Rhesus Monkey (98%), Bovine (94%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Lysophospholipases are enzymes that act on biological membranes to regulate the multifunctional lysophospholipids. The protein encoded by this gene hydrolyzes lysophosphatidylcholine in both monomeric and micellar forms. The use of alternate polyadenylation sites has been found for this gene. There are alternatively spliced transcript variants described for this gene but the full length nature is not known yet.Spezifikation
LYPLA1 | |
Polyclonal | |
Unconjugated | |
LYPLA1 | |
25KDL; Acyl-protein thioesterase 1; APT1; APT-1; CaIPLA2; calcium-independent phospholipase A2; calcium-independent phospholipase A2 isoform 1; calcium-independent phospholipase A2 isoform 2; hAPT1; hypothetical protein LOC550279; LPL1; LPL-I; Lypla1; lysophopholipase 1; lysophospholipase 1; Lysophospholipase I; lysophospholipid-specific lysophospholipase; LYSOPLA; lysoPLA I; phospholipase 1a; Pla1a; zgc:110260 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10434 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
O75608 | |
LYPLA1 | |
Recombinant fragment corresponding to a region within amino acids 1 and 230 of Human LYPLA1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |