missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAGEA4 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 55109-1-AP-150UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
The antibody is specific to MAGEA4.
This gene is a member of the MAGEA gene family. The members of this family encode proteins with 50 to 80% sequence identity to each other. The promoters and first exons of the MAGEA genes show considerable variability, suggesting that the existence of this gene family enables the same function to be expressed under different transcriptional controls. The MAGEA genes are clustered at chromosomal location Xq28. They have been implicated in some hereditary disorders, such as dyskeratosis congenita. At least four variants encoding the same protein have been found for this gene.Spezifikation
MAGEA4 | |
Polyclonal | |
Unconjugated | |
MAGEA4 | |
Cancer/testis antigen 1.4, CT1.4, MAGE 4 antigen, MAGE 41, MAGE 41 antigen, MAGE X2, MAGE X2 antigen, MAGE4, MAGE4A, MAGE4B, MAGEA4, melanoma antigen family A, 4, Melanoma associated antigen 4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
4103 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P43358 | |
MAGEA4 | |
Peptide | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |