Learn More
MAOA Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA527381
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: HepG2. Predicted reactivity: Mouse (89%), Rat (90%), Dog (90%), Pig (90%), Bovine (89%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes monoamine oxidase A, an enzyme that degrades amine neurotransmitters, such as dopamine, norepinephrine, and serotonin. The protein localizes to the mitochondrial outer membrane. The gene is adjacent to a related gene on the opposite strand of chromosome X. Mutation in this gene results in monoamine oxidase deficiency, or Brunner syndrome.Spezifikation
MAOA | |
Polyclonal | |
Unconjugated | |
MAOA | |
1110061B18Rik; AA407771; Amine oxidase [flavin-containing] A; HGNC:6833; Mao; MAOA; MAO-A; monoamine oxidase A; monoamine oxidase type A; RP1-201D17__B.2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
17161, 4128 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.33 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
P21397, Q64133 | |
MAOA | |
Recombinant fragment corresponding to a region within amino acids 166 and 513 of Human monoamine oxidase A | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |