missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAT2A Recombinant Rabbit Monoclonal Antibody (24GB530)
Rabbit Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA557074
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
MAT2A Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
The protein encoded by this gene catalyzes the production of S-adenosylmethionine (AdoMet) from methionine and ATP. AdoMet is the key methyl donor in cellular processes.
Spezifikation
| MAT2A | |
| Recombinant Monoclonal | |
| 0.6 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P18298, P31153, Q3THS6 | |
| MAT2A | |
| A synthesized peptide derived from human MAT2A (aa 50-138). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB530 | |
| Unconjugated | |
| MAT2A | |
| adoMet synthase 2; adoMet synthetase 2; AMS2; D630045P18Rik; MAT 2; MAT2A; MATA2; MATII; MAT-II; Methionine adenosyltransferase 2; methionine adenosyltransferase 2A; Methionine adenosyltransferase II; methionine adenosyltransferase II, alpha; non-hepatic-type S-adenosylmethionine synthetase; S-adenosylmethionine synthase; S-adenosylmethionine synthase isoform type-2; S-adenosylmethionine synthetase isoform type-2; Sams2; testicular tissue protein Li 121 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 171347, 232087, 4144 | |
| -20°C | |
| Liquid |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur