Learn More
STEMCELL Technologies MegaCult™-C Medium with Cytokines
Artikelnummer. 17118241
Encompass
Change view
Click to view available options
Menge:
24 x 2 ml
Packungsgröße:
24 Stück
Artikelnummer. Menge unitSize
17118241 24 x 2 ml 24 Stück
1 options
Artikelnummer. 17118241
Marke: STEMCELL Technologies 04901
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Medium with cytokines
  • MegaCult™-C Complete Kit with Cytokines includes all reagents required for the optimal growth of megakaryocyte progenitors (CFU-Mk) within double chamber slides and for the detection of CFU-Mk colonies by immunocytochemical staining
  • MegaCult™-C medium is optimized for the growth of CFU-Mk in human bone marrow, mobilized peripheral blood and cord blood samples
  • It is suitable for use with CD34+-enriched cells, mononuclear cells and cells isolated by other purification methods
Spezifikation
Zelltyp Hematopoietic Stem and Progenitor Cells
Zur Verwendung mit (Anwendung) Cell Culture, Colony Assay, Functional Assay
Menge 24 x 2 ml