Learn More
MedChemExpress Melanotan I
Artikelnummer. 16439298
Encompass_Preferred
Ansicht ändern
Click to view available options
Menge:
10 mg
Packungsgröße:
10 Milligramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
16439298 10 mg 10 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 16439298 Lieferant MedChemExpress Lieferanten-Nr. HYN2466
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Melanotan I is a potent non-selective melanocortin receptor (MCR) agonist. Melanotan I is a synthetic analogue of α-melanocyte stimulating hormone (α-MSH) that stimulates melanogenesis. Melanotan I can induce skin tanning by mimicking the actions of a-MSH on the melanocortin type 1 receptors (MC1R) of melanocytes. Melanotan I can be used for sunlight-induced skin cancers research.
Spezifikation
Reinheit 97.22%
CAS 75921-69-6
Inhalt und Lagerung Powder -80°C 2 years, -20°C 1 year∣The compound is unstable in solutions, freshly prepared is recommended.
Molekulargewicht 1646.85 g/mol
Produkttyp Peptides
Menge 10 mg
Versandbedingung Room temperature
Löslichkeit H2O : ≥ 50 mg/mL (30.36 mM)
Synonym MT-I∣ [Nle4,D-Phe7]-α-MSH