missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MEP1A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 24640-1-AP-20UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
MEP1A (Meprin A Subunit Alpha) is a Protein Coding gene. Among its related pathways are Collagen chain trimerization. Gene Ontology (GO) annotations related to this gene include metalloendopeptidase activity and metallopeptidase activity.Spezifikation
MEP1A | |
Polyclonal | |
Unconjugated | |
MEP1A | |
Endopeptidase 2, MEP1A, Meprin A subunit alpha, PABA peptide hydrolase, PPH alpha, PPHA | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
17287, 25684, 4224 | |
-20°C | |
Liquid |
Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P28825, Q16819, Q64230 | |
Mep1a | |
MEP1A Fusion Protein Ag18950 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |