missing translation for 'onlineSavingsMsg'
Learn More

Metapneumovirus Glycoprotein G Antibody - (HMPV/Homo sapiens/PER/CFI0466/2010/B), Novus Biologicals™

Rabbit Polyclonal Antibody

Marke:  Novus Biologicals NBP3-43769-100ul

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 30281488

  • 398.00 € / 100 Mikroliter

om dit product te kopen Registreer vandaag om een webaccount aan te maken

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Beschreibung

Beschreibung

Metapneumovirus Glycoprotein G Polyclonal antibody specifically detects Metapneumovirus Glycoprotein G in Virus samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Spezifikation

Spezifikation

Metapneumovirus Glycoprotein G
Polyclonal
ELISA 1:5000-1:10000
Rabbit
Antigen and protein A Affinity-purified
RUO
Virus
Purified
ELISA
Unconjugated
0.2 μm filtered solution in PBS
Produced in rabbits immunized with purified, recombinant Metapneumovirus Glycoprotein G (HMPV/Homo sapiens/PER/CFI0466/2010/B) (RefSeq#: AHV79646.1; Asp52-Ser238)
100 μL
Primary
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Aktionen

Aktionen

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.