Learn More
STEMCELL Technologies MethoCult™ H4435 Enriched
Artikelnummer. 17138221
Encompass
Change view
Click to view available options
Menge:
100 ml
Packungsgröße:
100 Milliliter
Artikelnummer. Menge unitSize
17138221 100 ml 100 Milliliter
1 options
Artikelnummer. 17138221
Marke: STEMCELL Technologies 04435
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Methylcellulose-based medium with recombinant cytokines for human cells
  • MethoCult™ H4435 Enriched (MethoCult™ GF+ H4435) is a complete methylcellulose-based medium for the growth and enumeration of hematopoietic progenitor cells in colony-forming unit (CFU) assays of human bone marrow, mobilized peripheral blood, peripheral blood, and cord blood samples
  • This medium supports optimal growth of erythroid progenitor cells (BFU-E and CFU-E), granulocyte-macrophage progenitor cells (CFU-GM, CFU-G and CFU-M), and multipotential granulocyte, erythroid, macrophage and megakaryocyte progenitor cells (CFU-GEMM)
  • MethoCult™ H4435 Enriched is recommended for use with CD34+ cells and other purified cell populations, and for CFU assays performed as part of LTC-IC assays
Spezifikation
Zelltyp Hematopoietic Stem and Progenitor Cells
Zur Verwendung mit (Anwendung) Cell Culture, Colony Assay, Functional Assay
Menge 100 ml