missing translation for 'onlineSavingsMsg'
Learn More

Methyl4'-Hydroxybiphenyl-3-Carboxylat, 95%, Thermo Scientific™

Catalog Number 15468217
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
192376-76-4
C14H12O3
228.247
MFCD06409246
ZWSVMMHYHQNTKA-UHFFFAOYSA-N
methyl 3-4-hydroxyphenyl benzoate, methyl 4'-hydroxy-1,1'-biphenyl-3-carboxylate, 4'-hydroxybiphenyl-3-carboxylic acid methyl ester, 4-3-methoxycarbonylphenyl phenol, amtda024, acmc-1c4b1, 3'-methoxycarbonylbiphenyl-4-ol, 4-3-methoxycarbonyl phenyl phenol, 4'-hydroxy-3-methoxycarbonyl biphenyl, methyl 4'-hydroxybiphenyl-3-carboxylate
5097337
Methyl-3-(4-hydroxyphenyl)benzoat
COC(=O)C1=CC=CC(=C1)C2=CC=C(C=C2)O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
192376-76-4
C14H12O3
228.247
MFCD06409246
ZWSVMMHYHQNTKA-UHFFFAOYSA-N
methyl 3-4-hydroxyphenyl benzoate, methyl 4'-hydroxy-1,1'-biphenyl-3-carboxylate, 4'-hydroxybiphenyl-3-carboxylic acid methyl ester, 4-3-methoxycarbonylphenyl phenol, amtda024, acmc-1c4b1, 3'-methoxycarbonylbiphenyl-4-ol, 4-3-methoxycarbonyl phenyl phenol, 4'-hydroxy-3-methoxycarbonyl biphenyl, methyl 4'-hydroxybiphenyl-3-carboxylate
5097337
Methyl-3-(4-hydroxyphenyl)benzoat
COC(=O)C1=CC=CC(=C1)C2=CC=C(C=C2)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material Methyl 4'-hydroxybiphenyl-3-carboxylate
Menge 1 g
UN-Nummer UN3077
Formelmasse 228.25
Reinheit (%) 95%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'