Learn More
Methyloctanoat, 99 %, Thermo Scientific Chemicals
Catalog Number p-7077833
Click to view available options
Menge:
1 g
5 g
Methyl octanoate
111-11-5
0.8780g/mL
79.0°C (12.0 mmHg)
73°C
Echt
98.5% min. (GC)
C9H18O2
CH3(CH2)6COOCH3
1.4165 to 1.4185
0.878
methyl caprylate, caprylic acid methyl ester, methyl n-octanoate, octanoic acid, methyl ester, uniphat a20, octanoic acid methyl ester, methyl octylate, methyl caprylate natural, unii-7mo740x6ql, fema no. 2728
Solubility in water: insoluble.
JGHZJRVDZXSNKQ-UHFFFAOYSA-N
CCCCCCCC(=O)OC
Methyloctanoat
158.24
8091
CHEBI:87432
158.24
99%
Methyl octanoate
111-11-5
0.8780g/mL
79.0°C (12.0 mmHg)
73°C
Echt
98.5% min. (GC)
C9H18O2
CH3(CH2)6COOCH3
1.4165 to 1.4185
0.878
methyl caprylate, caprylic acid methyl ester, methyl n-octanoate, octanoic acid, methyl ester, uniphat a20, octanoic acid methyl ester, methyl octylate, methyl caprylate natural, unii-7mo740x6ql, fema no. 2728
Solubility in water: insoluble.
JGHZJRVDZXSNKQ-UHFFFAOYSA-N
CCCCCCCC(=O)OC
Methyloctanoat
158.24
8091
CHEBI:87432
158.24
99%
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Verpackung Glasflasche
Menge 1 g