Learn More
MGP Mouse anti-Human, Mouse, Clone: 1A1C3, Proteintech
Mouse Monoclonal Antibody
Marke:  Proteintech 60055-1-IG-20UL
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 16897005
  • 145.00 € / 20 Mikroliter
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Beschreibung
Beschreibung
MGP is associates with the organic matrix of bone and cartilage. MGP is thought to act as an inhibitor of bone formation.
Spezifikation
Spezifikation
MGP
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P08493, P19788
Mgp
MGP Fusion Protein Ag1091
20 μL
Primary
Human, Mouse
Antibody
IgG2a
Immunohistochemistry (Paraffin), Western Blot
1A1C3
Unconjugated
MGP
GIG36, matrix Gla protein, MGLAP, MGP, NTI
Mouse
Protein A
RUO
17313, 4256
-20°C