Learn More
MGP Mouse anti-Human, Mouse, Clone: 1A1C3, Proteintech
Mouse Monoclonal Antibody
145.00 € - 410.00 €
Spezifikation
| Antigen | MGP |
|---|---|
| Klon | 1A1C3 |
| Konzentration | 1 mg/mL |
| Anwendungen | Immunohistochemistry (Paraffin), Western Blot |
| Klassifikation | Monoclonal |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
16897005
|
Proteintech
60055-1-IG-20UL |
20 μL |
145.00 €
20 Mikroliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
|
16887005
|
Proteintech
60055-1-IG-150UL |
150 μL |
410.00 €
150 Mikroliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
MGP is associates with the organic matrix of bone and cartilage. MGP is thought to act as an inhibitor of bone formation.Spezifikation
| MGP | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| GIG36, matrix Gla protein, MGLAP, MGP, NTI | |
| Mgp | |
| IgG2a | |
| Protein A | |
| Antibody |
| 1A1C3 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Mouse | |
| P08493, P19788 | |
| 17313, 4256 | |
| MGP Fusion Protein Ag1091 | |
| Primary | |
| -20°C | |
| MGP |