Learn More
Ibidi micro-Insert 4 Well in μ-Dish 35 mm, High ibiTreat
Artikelnummer. 16481661
Encompass_Preferred
Ansicht ändern
Click to view available options
Anzahl Wells:
4
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Anzahl Wells
16481661 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 16481661 Lieferant Ibidi Lieferanten-Nr. 80406
Please to purchase this item. Need a web account? Register with us today!
micro-Insert 4 Well in μ-Dish 35 mm, High ibiTreat is a 4 well silicone insert for live cell imaging.
  • A 4 well silicone insert for live cell imaging of both adherent and suspension cells
  • in a μ-Dish 35 mm high with #1.5 Polymer Coverslip bottom
  • tissue culture treated
  • sterilized
Spezifikation
Höhe (metrisch) 35 mm
Menge 30
Sterilität Steril
Anzahl Wells 4