Learn More
Ibidi micro-Insert 4 Well FulTrac in μ-Dish 35 mm, High ibiTreat
Artikelnummer. 17276183
Encompass_Preferred
Ansicht ändern
Click to view available options
Anzahl Wells:
4
Packungsgröße:
30 Stück
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Anzahl Wells unitSize
17276183 4 30 Stück
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 17276183 Lieferant Ibidi Lieferanten-Nr. 80486
Nur noch null auf Lager
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
micro-Insert 4 Well FulTrac in μ-Dish 35 mm, High ibiTreat is a 4 well silicone insert for live cell imaging.
  • A 4 well silicone insert used for tracing individual cells (adherent and suspension)
  • in a μ-Dish 35 mm high with #1.5 Polymer Coverslip bottom
  • tissue culture treated
  • sterilized
Spezifikation
Höhe (metrisch) 35 mm
Menge 30
Sterilität Steril
Anzahl Wells 4