Learn More
Kern Sohn Mikroskopkamera ODC
Artikelnummer. 17319541
Encompass_Preferred
Click to view available options
Anschlusstyp:
USB 2.0
USB 3.0
Auflösung:
20 MP
5,1 MP
Größe:
1/2.5 Zoll Sensor
1 Zoll Sensor
Kompakt, 1 Zoll Sensor
Packungsgröße:
1 Stück
Artikelnummer. 17319541
Marke: Kern Sohn ODC 832
Nur noch null auf Lager
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Specialists in microscopy for measurement, counting, documentation, archiving and image processing.
  • The universal microscope cameras can be used anywhere and can be connected to the microscope as well as to a laptop or PC using the USB cable (USB 2.0)
  • Through the proven CMOS technology, in connection with the USB 2.0 the images are shown quickly and clearly
  • These cameras are also ideal for more demanding applications, such as, for example, darkfield, phase contrast and for fluorescence applications.
  • Your daily work is made significantly easier with the very best synchronisation, a high frame rate as well as stable image performance together with our Microscope VIS software which we deliver with the product.
  • The power supply is through the USB cable, which means that no additional power supply is required
  • As well as the camera, the delivery includes our multi-lingual KERN Microscope VIS Basic KERN OXM 901, a USB cable (Length: 2000 mm) various eyepiece adapters and an object micrometer to calibrate the software.
  • Please order the appropriate C-mount adapter to fit your KERN microscope now.
Spezifikation
Zur Verwendung mit (Geräte) Mikroskope
Anschlusstyp USB 3.0
Auflösung 5,1 MP
Arten von Sensoren CMOS
Größe 1/2.5 Zoll Sensor