missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MMP-9 Antibody (S51-82), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
Marke: Novus Biologicals NBP2-59699-25ug
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
MMP-9 Monoclonal specifically detects MMP-9 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
Spezifikation
| MMP-9 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:100 | |
| 92 kDa gelatinase, 92 kDa type IV collagenase, CLG4B, EC 3.4.24, EC 3.4.24.35, Gelatinase B, GELB, macrophage gelatinase, MANDP2, matrix metallopeptidase 9, matrix metalloproteinase 9, matrix metalloproteinase-9, MMP-9, type V collagenase | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Detecs ∼92kDa and ∼82kDa (pro and active forms). | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG2a |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| S51-82 | |
| Unconjugated | |
| PBS (pH 7.4), 50% glycerol with 0.09% Sodium Azide | |
| MMP9 | |
| Fusion protein amino acids 1-708 (full length) of rat MMP9 | |
| 25 μg | |
| Angiogenesis, Cancer, Cellular Markers, Extracellular Matrix, GPCR | |
| 4318 | |
| Human, Mouse, Rat | |
| Purified |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only (RUO)
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur