missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MMP3 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 1F5A9, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech 66338-1-IG-150UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
MMP3 (stromelysin-1) belongs to the peptidase M10A subfamily. It is secreted in its pro-protein form, and is activated by extracellular proteinase cleavage. MMP3 is capable of binding to zinc and calcium ions, and has metallo-endopeptidase activity. The active enzyme degrades collagen, gelatin, fibronectin, and laminin. It is involved in normal cellular processes such as wound repair and tissue remodeling. Mutations in the gene can result in coronary heart disease 6.Spezifikation
| MMP3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P03957, P08254, P28862 | |
| Mmp3 | |
| MMP3 Fusion Protein Ag12359 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Porcine, Rat | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1F5A9 | |
| Unconjugated | |
| MMP3 | |
| CHDS6, Matrix metalloproteinase 3, MMP 3, MMP3, SL 1, STMY, STMY1, STR1, Stromelysin 1, Transin 1 | |
| Mouse | |
| Protein G | |
| RUO | |
| 171045, 17392, 396769, 4314 | |
| -20°C | |
| Liquid |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur