missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Morphine Polyclonal Antibody, Invitrogen™
Sheep Polyclonal Antibody
Marke: Invitrogen PA175296
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
PA1-75296 specifically detects Morphine. PA1-75296 has been successfully used in ELISA applications. The PA1-75296 immunogen is Morphine(3)-BSA.
Morphine Base is an opiate alkaloid isolated from the plant Papaver somniferum and produced synthetically. Morphine binds to and activates specific opiate receptors (delta, mu and kappa), each of which are involved in controlling different brain functions. In the central nervous and gastrointestinal systems, this agent exhibits widespread effects including analgesia, anxiolysis, euphoria, sedation, respiratory depression, and gastrointestinal system smooth muscle contraction. [National Center for Biotechnology Information. PubChem Compound Database; CID=5288826].Spezifikation
Morphine | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Sheep | |
purified | |
RUO | |
Chemical | |
Antibody | |
IgG |
ELISA | |
Unconjugated | |
C17H19NO3; Morphia; Morphin; Morphinum; Morphium | |
Morphine(3)-BSA | |
1 mg | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid | |
Chemical |