Learn More
MOS Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA559173
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: CKISDFGCSE KLEDLLCFQT PSYPLGGTYT HRAPELLKGE GVTPKADIYS FAITLWQMTT KQAPYSGERQ HILYAVVAYD LRPSLSAAVF Highest antigen sequence identity to the following orthologs: Mouse - 72%, Rat - 73%.
Protein kinases are enzymes that transfer a phosphate group from a phosphate donor, generally the g phosphate of ATP, onto an acceptor amino acid in a substrate protein. By this basic mechanism, protein kinases mediate most of the signal transduction in eukaryotic cells, regulating cellular metabolism, transcription, cell cycle progression, cytoskeletal rearrangement and cell movement, apoptosis, and differentiation. With more than 500 gene products, the protein kinase family is one of the largest families of proteins in eukaryotes. The family has been classified in 8 major groups based on sequence comparison of their tyrosine (PTK) or serine/threonine (STK) kinase catalytic domains. The STE group (homologs of yeast Sterile 7, 11, 20 kinases) consists of 50 kinases related to the mitogen-activated protein kinase (MAPK) cascade families (Ste7/MAP2K, Ste11/MAP3K, and Ste20/MAP4K). MAP kinase cascades, consisting of a MAPK and one or more upstream regulatory kinases (MAPKKs) have been best characterized in the yeast pheromone response pathway. Pheromones bind to Ste cell surface receptors and activate yeast MAPK pathway.Spezifikation
MOS | |
Polyclonal | |
Unconjugated | |
MOS | |
C-MOS; c-mos oncogene; c-mos proto-oncogene; MGC119962; MGC119963; Moloney sarcoma oncogene; mos; MOS proto-oncogene, serine/threonine kinase; MOS proto-oncogene, serine/threonine kinase L homeolog; mos.L; mosxe; MSV; oncogene MOS, Moloney murine sarcoma virus; oocyte maturation factor mos; p39; p39-mos; pp39-mos; protein kinase; proto-oncogene c-Mos; Proto-oncogene serine/threonine-protein kinase mos; serine/threonine-protein kinase mos; v-mos Moloney murine sarcoma viral oncogene homolog; v-mos Moloney murine sarcoma viral oncogene homolog L homeolog; XELAEV_18032032mg | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4342 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P00540 | |
MOS | |
Recombinant protein corresponding to Human MOS | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only