Learn More
Ibidi Mounting Medium with DAPI
Beschreibung
- A mounting medium with DAPI that is optimized for fluorescence microscopy with Ibidi μ-Slides and μ-Dishes
- non-drying
- non-hardening
- volume stable
- 15 mL
Spezifikation
Spezifikation
| Menge | 1 |
| Produkttyp | Eindeckmedium |
| Volumen (metrisch) | 5 mL |