Learn More
Mouse anti-Human IgG4 Secondary Antibody, HRP, Invitrogen™
Mouse Monoclonal Secondary Antibody
Marke: Invitrogen MA134437
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
There is no cross reaction with human IgG1, IgG2 or IgG3 isotypes. A suggested positive control for this product is tonsil. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Spezifikation
Human IgG4 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 1% BSA and 0.002% Bromonitrodioxane, 0.002% Methylisothiazolone, 0.05% ProClin 300 | |
Purified IgG4 | |
100 μg | |
Secondary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG1 |
ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) | |
HP6025 | |
HRP | |
Mouse | |
Protein A | |
RUO | |
Human | |
Liquid |