Learn More
ACROBiosystems Mouse CD155 / PVR Protein, Mouse IgG2a Fc Tag
423.38 € - 4306.37 €
Spezifikation
Zugriffsnummer | NP_081790.1 |
---|---|
Molekulargewicht | 61.8 kDa |
Quelle | Mouse CD155, Mouse IgG2a Fc Tag (CD5-M5254) is expressed from human 293 cells (HEK293). It contains AA Asp 29 - Leu 348 (Accession # Q8K094-1). |
Immunogen | Asp 29 - Leu 348 |
Lagerungsbedingungen | -20°C |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16457309
|
ACROBiosystems
CD5-M5254-1MG |
1 mg |
4306.37 €
1 Milligramm |
Verfügbar ab: 07-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
15836217
|
ACROBiosystems
CD5-M5254-50UG |
50 μg |
423.38 €
50 Mikrogramm |
Verfügbar ab: 07-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
CD155 (cluster of differentiation 155) also known as the poliovirus receptor is a protein that is encoded by the PVR gene. CD155 is a Type I transmembrane glycoprotein in the immunoglobulin superfamily. Commonly known as Poliovirus Receptor (PVR) due to its involvement in the cellular poliovirus infection in primates, CD155's normal cellular function is in the establishment of intercellular adherens junctions between epithelial cells. The role of CD155 in the immune system is unclear, though it may be involved in intestinal humoral immune responses. Subsequent data has also suggested that CD155 may also be used to positively select MHC-independent T cells in the thymus.
This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 61.8 kDa. The protein migrates as 35 kDa and 70-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_081790.1 | |
Mouse CD155, Mouse IgG2a Fc Tag (CD5-M5254) is expressed from human 293 cells (HEK293). It contains AA Asp 29 - Leu 348 (Accession # Q8K094-1). | |
-20°C | |
CD155 | |
Mouse | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Immune Checkpoint Proteins |
61.8 kDa | |
Asp 29 - Leu 348 | |
PVR,FLJ25946,PVS,CD155,TAGE4,HVED,NECL5 | |
Unconjugated | |
Mouse IgG Fc Tag | |
Less than 0.1 EU / μg by the LAL method. | |
Powder | |
>95% as determined by reduced SDS-PAGE |