Learn More
Invitrogen™ CXCL15 Mouse ELISA Kit
Sandwich ELISA Kit
Marke: Invitrogen™ EMCXCL15
Weitere Details : Netto-Gewicht : 0.00400kg
Beschreibung
The Mouse CXCL15 ELISA quantitates Ms CXCL15 in mouse serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Ms CXCL15. Principle of the method The Mouse CXCL15 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Chemotaktisch gegenüber Neutrophilen. Beteiligt am lungenspezifischen Neutrophilentransport unter normalen und entzündlichen Bedingungen.Spezifikation
Gehäuse Q9WVL7 | |
0,8 ng/mL | |
Festphasen-Sandwich-ELISA | |
ELISA-Kit | |
Plasma, Serum, Überstand | |
96 Assays | |
ELISA, Protein-Assays, Proteinbiologie, Krebsbiologie, Immunologie, Neurowissenschaften, Neurobiologie, Proteinnachweis, Proteinanalyse | |
20309 | |
< 12 % | |
Vorbeschichtete 96-Well-Platte, Assay-Verdünnungskonzentrat, Biotinylierter Nachweisantikörper, Waschpuffer, Stopplösung, Dichtungsfolien für Mikrotiterplatten, Chromogen, Stopplösung, Dichtungsfolien für Mikrotiterplatten | |
96 Test-Kit | |
Entzündungen | |
2 bis 8 °C | |
Maus | |
4 Std 45 Min |
0.819 bis 200 ng/ml | |
0.8 bis 200 ng/ml | |
Biotin | |
Chemokin und Rezeptoren | |
Dieses ELISA-Antikörperpaar erkennt Maus-CXCL15. Andere Arten wurden nicht bestimmt. | |
Maus | |
Kolorimetrisches Mikrotiterplatten-Lesegerät | |
Il8, Scyb15, lungkine,weche | |
< 10 % | |
HRP | |
RUO | |
Plasma, 50 μl; Serum, 50 μl; Überstand, 100 μl | |
Il8, Scyb15, lungkine, weche | |
1 Std 20 Min |