missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse EDAR Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Marke: R&D Systems AF745
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
EDAR Polyclonal specifically detects EDAR in Mouse samples. It is validated for Western Blot, Blockade of Receptor-ligand Interaction.
Spezifikation
| EDAR | |
| Polyclonal | |
| Unconjugated | |
| Q9R187 | |
| EDAR | |
| Mouse myeloma cell line NS0-derived recombinant mouse EDAR Glu27-Ala187 Accession # Q9R187 | |
| 100 μg | |
| Primary | |
| Detects mouse EDAR in direct ELISAs and Western blots. | |
| Mouse | |
| IgG |
| Western Blot, Blockade of Receptor-ligand Interaction | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Blockade of Receptor-ligand Interaction | |
| Anhidrotic ectodysplasin receptor 1, DLED3, Ectodermal dysplasia receptor, ectodysplasin 1, anhidrotic receptor, ectodysplasin A receptor, Ectodysplasin-A receptor, ED1R, ED5, EDA1R, EDA3EDA-A1 receptor, EDA-A1R, Edar, FLJ94390, HRM1, mouse, homolog of, tumor necrosis factor receptor superfamily member EDAR | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 10913 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur