missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Fc gamma RIII (CD16) APC-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 4 publications
218.00 € - 491.00 €
Spezifikation
| Antigen | Fc gamma RIII (CD16) |
|---|---|
| Klon | 275003 |
| Verdünnung | Flow Cytometry 10 uL/10^6 cells |
| Anwendungen | Flow Cytometry |
| Klassifikation | Monoclonal |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
18711028
![]() |
R&D Systems
FAB19601A-025 |
25 Tests |
218.00 €
25 Tests |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
18738216
![]() |
R&D Systems
FAB19601A-100 |
100 Tests |
491.00 €
100 Tests |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
Fc gamma RIII (CD16) Monoclonal specifically detects Fc gamma RIII (CD16) in Mouse samples. It is validated for Flow Cytometry.Spezifikation
| Fc gamma RIII (CD16) | |
| Flow Cytometry 10 uL/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| CD16, CD16A, Fc fragment of IgG receptor IIIa, FCG3, FCGR3, FCGRIII, FCR-10, FCRIII, FCRIIIA, IGFR3, IMD20 | |
| Fcgr3 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Detects mouse Fc gamma RIIIA/B (CD16) in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse Fcrl3/CD16-2, rmFc gamma RIA or rhFc gamma RIIA is observed. |
| 275003 | |
| Flow Cytometry | |
| APC | |
| Rat | |
| Mouse | |
| Q5D5J5 | |
| 2214 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Fc gamma RIII Ala31-Thr215 Accession # Q5D5J5 | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts