Learn More
ACROBiosystems Mouse HMGB1 Protein, His Tag
483.87 € - 3495.90 €
Spezifikation
Zugriffsnummer | NP_034569.1 |
---|---|
Molekulargewicht | 26.8 kDa |
Quelle | Mouse HMGB1, His Tag (HM1-M52H6) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Glu 215 (Accession # P63158-1). |
Immunogen | Met 1 - Glu 215 |
Lagerungsbedingungen | -20°C |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16447639
|
ACROBiosystems
HM1-M52H6-1MG |
1 mg |
3495.90 €
1 Milligramm |
Verfügbar ab: 07-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
16437639
|
ACROBiosystems
HM1-M52H6-100UG |
100 μg |
483.87 €
100 Mikrogramm |
Verfügbar ab: 07-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
High-mobility group protein B1 (HMGB1) is also known as high-mobility group protein 1 (HMG-1) and amphoterin, is a member of the HMGB family consisting of three members, HMGB1, HMGB2 and HMGB3. HMGB1 is a non-histone architectural chromosomal protein ubiquitously present in all vertebrate nuclei and binds double-stranded DNA without sequence specificity. The mechanism of inflammation and damage is binding to TLR4, which mediates HMGB1-dependent activation of macrophage cytokine release. This positions HMGB1 at the intersection of sterile and infectious inflammatory responses. HMGB1 has been studied as a DNA vaccine adjuvant and a target for cancer therapy.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 26.8 kDa. The protein migrates as 33 kDa and 35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_034569.1 | |
Mouse HMGB1, His Tag (HM1-M52H6) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Glu 215 (Accession # P63158-1). | |
-20°C | |
HMGB1 | |
Mouse | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |
26.8 kDa | |
Met 1 - Glu 215 | |
HMGB1,HMG1,HMG3,SBP-1 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |