missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse ICAM-5 Antibody, R&D Systems™

Rat Monoclonal Antibody
176.00 € - 496.00 €
Spezifikation
| Antigen | ICAM-5 |
|---|---|
| Klon | 191321 |
| Konzentration | LYOPH |
| Verdünnung | Western Blot 1 ug/mL |
| Anwendungen | Western Blot |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
18709427
![]() |
R&D Systems
MAB1173-SP |
25 μg |
176.00 €
25 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
18166131
![]() |
R&D Systems
MAB1173 |
500 μg |
496.00 €
500 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
ICAM-5 Monoclonal specifically detects ICAM-5 in Mouse samples. It is validated for Western Blot.Spezifikation
| ICAM-5 | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| Rat | |
| Mouse | |
| Q60625 | |
| 7087 | |
| Mouse myeloma cell line NS0-derived recombinant mouse ICAM-5 Leu31-Arg828 (Pro47Arg) Accession # Q60625 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse ICAM-5 in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant mouse (rm) DCC, rmICAM-1, rhICAM-1, rmICAM-2, rhICAM-3, rmMadCAM, rhCD31, or rmVCAM-1. |
| 191321 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ICAM5, ICAM-5, intercellular adhesion molecule 5, intercellular adhesion molecule 5, telencephalin, Telencephalin, TLCNTelencephalin, TLNtelencephalin | |
| ICAM5 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts