Learn More
ACROBiosystems Mouse IGF-I R / CD221 Protein, His Tag
423.38 € - 3108.82 €
Spezifikation
Zugriffsnummer | NP_034643.2 |
---|---|
Molekulargewicht | 81.3 kDa, 24.3 kDa |
Quelle | Mouse IGF-I R, His Tag (IGR-M5223) is expressed from human 293 cells (HEK293). It contains AA Glu 31 - His 936 (Accession # Q60751-1). |
Immunogen | Glu 31 - His 936 |
Lagerungsbedingungen | -20°C |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16467669
|
ACROBiosystems
IGR-M5223-1MG |
1 mg |
3108.82 €
1 Milligramm |
Verfügbar ab: 24-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
16457669
|
ACROBiosystems
IGR-M5223-100UG |
100 μg |
423.38 €
100 Mikrogramm |
Verfügbar ab: 24-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
The Insulin-like Growth Factor 1 Receptor (IGF1) is also known as CD221, JTK13. and is a transmembrane receptor that is activated by IGF-1 and by the related growth factor IGF-2. It belongs to the large class of tyrosine kinase receptors. This receptor mediates the effects of IGF-1, which is a polypeptide protein hormone similar in molecular structure to insulin. IGF1R is make up of two alpha subunits and two beta subunits ,the Both the α and β subunits are synthesized from a single mRNA precursor. The precursor is then glycosylated, proteolytically cleaved, and crosslinked by cysteine bonds to form a functional transmembrane αβ chain.The α chains are located extracellularly while the β subunit spans the membrane and are responsible for intracellular signal transduction upon ligand stimulation. IGF1R have a binding site for ATP, which is used to provide the phosphates for autophosphorylation. There is a 60% homology between IGF1R and the insulin receptor. In response to ligand binding, the α chains induce the tyrosine autophosphorylation of the β chains. This event triggers a cascade of intracellular signaling that, while somewhat cell type specific, often promotes cell survival and cell proliferation.
This protein carries a polyhistidine tag at the C-terminus.This protein contains a furin convertase cleavage site, 738-RRRR-741, and will be partially processed into N (α chain) and C-terminal fragment (partial β chain) with calculated MW of 81.3 kDa andSpezifikation
NP_034643.2 | |
Mouse IGF-I R, His Tag (IGR-M5223) is expressed from human 293 cells (HEK293). It contains AA Glu 31 - His 936 (Accession # Q60751-1). | |
-20°C | |
IGF-I R | |
Mouse | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |
81.3 kDa, 24.3 kDa | |
Glu 31 - His 936 | |
IGF1R,IGFR,JTK13,CD221,MGC142170,MGC142172,MGC18216 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |