Learn More
Mouse IgG1 Isotype Control, PE, Invitrogen™
Mouse Isotype Control
Marke: Invitrogen MA518097
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA5-18097 is negative on all human cells and cell lines tested. This antibody recognizes a rat cell surface marker, and therefore cannot be used as a negative control in this species. Test results have shown that MA5-18097 is also suitable for use as a negative control with bovine, ovine, porcine, equine, canine, lapine and guinea-pig tissues. For FACS analysis, use 10 µL of the suggested working dilution to label 1x10^6 cells in 100 µL. Reconstitute with 1 mL distilled water. After reconstitution, store product undiluted at 4oC in the dark.
Mouse IgG1 is a subtype of immunoglobulin G antibody that is produced by B cells in response to an antigen. It is the most abundant IgG subtype in the serum and plays a crucial role in the adaptive immune response. Mouse IgG1 is composed of two heavy chains and two light chains, and it is involved in opsonization, complement activation, and antibody-dependent cell-mediated cytotoxicity. It is also involved in the regulation of immune responses, including the activation of T cells and the inhibition of B cell activation. Mouse IgG1 is widely used in research and diagnostic applications, including ELISA, Western blotting, and immunohistochemistry. It is also used in therapeutic applications, such as immunotherapy and vaccine development.Spezifikation
Mouse IgG1 | |
Conc. Not Determined | |
PBS with 1% BSA, 5% sucrose and 0.09% sodium azide | |
Mouse | |
Protein A | |
RUO | |
Not Applicable | |
Lyophilized |
Control, Flow Cytometry | |
PE | |
IgG1; Igh-4; Ighg1; Immunoglobulin G; Immunoglobulin G1; immunoglobulin heavy constant gamma 1 (G1m marker); ImmunoglobulinG; ImmunoglobulinG1; VH7183 | |
Rat thymocyte membrane glycoproteins | |
100 Tests | |
Isotype Control (Primary) | |
4° C, store in dark, DO NOT FREEZE! | |
IgG1 |