missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mouse IgM Isotype Control, Invitrogen™
Mouse Isotype Control
Marke: Invitrogen™ MGM00
Weitere Details : Netto-Gewicht : 0.10500kg
Beschreibung
Mouse IgM Isotype Control for Flow
Mouse IgM Control antibody specifically detects Mouse IgM in Not Applicable samples. It is validated for Flow Cytometry.Spezifikation
Mouse IgM | |
Monoclonal | |
Unconjugated | |
PBS with 0.1% sodium azide | |
Mouse | |
500 μL | |
Whole Antibody | |
1 Each | |
Not Applicable | |
Liquid | |
50 Tests |
Flow Cytometry | |
0.1mg/mL | |
Mouse IgM Antibody, 0.5mL, 0.1mg/mL Concentration, Mouse Host, Whole Antibody Fragment, 0.2μm Filtration, Unconjugated, IgM Isotype, Used For Cell Analysis, Flow Cytometry Antibodies And Secondary Detection | |
AGM1; AI326478; Igh6; Igh-6; Ighm; Igh-M; Igm; immunoglobulin heavy constant mu; MU; muH; VH | |
purified | |
ASR | |
Primary | |
0.1% Sodium azide | |
2-8°C | |
IgM | |
Wet Ice |
Analyte Specific Reagent. Analytical and performance characteristics are not established.