missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse IL-15 DuoSet, R&D Systems™
Mouse IL-15 DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
N.V.
Spezifikation
Assay-Bereich | 125.00 to 8000 pg/mL |
---|---|
Kreuzreaktivität | <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules. |
Gen-ID (Entrez) | 16168 |
Produkttyp | ELISA Development Kit |
Konjugat | Biotin |
Beschreibung
The Mouse IL-15 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies mouse IL-15.Spezifikation
125.00 to 8000 pg/mL | |
16168 | |
Biotin |
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules. | |
ELISA Development Kit | |
Mouse |