missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mouse IFN-gamma Recombinant Protein, eBioscience™
Recombinant Protein
Marke: Invitrogen™ BMS326
Weitere Details : Netto-Gewicht : 0.09500kg
Beschreibung
IFN gamma (Interferon gamma, Type II interferon) is a macrophage activation factor, and immune interferon that is produced primarily by T-lymphocytes and natural killer cells in response to antigens, mitogens, Staphylococcus enterotoxin B, phytohemaglutanin and other cytokines. IFN gamma is a dimeric protein consisting of two 146 amino acid subunits. IFN gamma is a glycoprotein that exists, functionally, as a homodimer of approximately 45 kDa. On SDS-PAGE, IFN gamma appears as a combination of 25, 20 and minor 15.5 kDa bands as a result of differential glycosylation. The biological activity of the IFN gamma homodimer is highly species specific. Human IFN gamma does not show cross-reactivity with mouse. IFN gamma function includes the following: antiviral activity, tumor antiproliferative activity, induction of class I and II MHC, macrophage activation, and enhanced immunoglobulin secretion by B lymphocytes. IFN gamma is involved in cytokine regulation and also acts synergistically with other cytokines. Activation of IFN gamma takes place through binding of IFN gamma receptor I and II, and activating the JAK-STAT pathway. IFN gamma does not show any homology with IFN alpha or IFN beta but human IFN gamma shows about 40% sequence homology with mouse IFN gamma. IFN gamma is upregulated by IL2, FGF basic, EGF and downregulated by vitamin D3 or DMN. IFN gamma gene mutations are associated with aplastic anemia. Description: Mouse IFN-gamma is a regulatory protein produced by activated NK cells and CD4+TCRalpha beta, CD8+TCR alpha beta, and TCR gamma lambda T cells. The biological activities of IFN-gamma include the specific binding to a single class of high affinity receptors. Recombinant mouse IFN-gamma is a 15.6 kDa protein containing 134 amino acid residues. Purity: >95%. Molecular Weight: 15.6 kDa.Spezifikation
P01580 | |
PBS with no preservative; pH 7.4 | |
15.6kDa | |
7.4 | |
E. coli | |
RUO | |
Mouse IFN gamma | |
The ED50 was determined by a cytopathic effect inhibition assay with mouse NIH 3T3 cells challenged with EMC virus is <0.1ng/mL, corresponding to a specific activity of >1 x 107 units/mg. | |
Mouse | |
Ms | |
Recombinant | |
Recombinant |
Control | |
15978 | |
Mouse IFN gamma | |
100μg | |
-20°C | |
E. Coli | |
Ifng | |
Recombinant Protein | |
Unconjugated | |
E. coli | |
Lyophilized | |
Affinity chromatography |