Learn More
ACROBiosystems Mouse LILRB4 / CD85k / ILT3 Protein, His Tag
544.34 € - 2927.37 €
Spezifikation
Zugriffsnummer | NP_038560.1 |
---|---|
Molekulargewicht | 25.9 kDa |
Quelle | Mouse LILRB4, His Tag (CDK-M5229) is expressed from human 293 cells (HEK293). It contains AA Gly 24 - Lys 238 (Accession # Q64281-1). |
Immunogen | Gly 24 - Lys 238 |
Lagerungsbedingungen | -20°C |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16487379
|
ACROBiosystems
CDK-M5229-1MG |
1 mg |
2927.37 €
1 Milligramm |
Verfügbar ab: 21-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
15836417
|
ACROBiosystems
CDK-M5229-100UG |
100 μg |
544.34 €
100 Mikrogramm |
Verfügbar ab: 21-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
Leukocyte immunoglobulin-like receptor subfamily B member 4 (LILRB4) is also known as CD85 antigen-like family member K (CD85K), Immunoglobulin-like transcript 3 (ILT-3), Leukocyte immunoglobulin-like receptor 5 (LIR-5), Monocyte inhibitory receptor HM18, which belongs to the leukocyte immunoglobulin-like receptor (LIR) family. LILRB4 / CD85K contains 2 Ig-like C2-type (immunoglobulin-like) domains. CD85K is detected in monocytes, macrophages, dendritic cells, lung, natural killer cells and B-cells. LILRB4 / CD85K is receptor for class I MHC antigens. CD85K recognizes a broad spectrum of HLA-A, HLA-B, HLA-C and HLA-G alleles, involved in the down-regulation of the immune response and the development of tolerance. LILRB4 interferes with TNFRSF5-signaling and NF-kappa-B up-regulation and inhibits receptor-mediated phosphorylation of cellular proteins and mobilization of intracellular calcium ions.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 25.9 kDa. The protein migrates as 33-42 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_038560.1 | |
Mouse LILRB4, His Tag (CDK-M5229) is expressed from human 293 cells (HEK293). It contains AA Gly 24 - Lys 238 (Accession # Q64281-1). | |
-20°C | |
LILRB4 | |
Mouse | |
PBS, pH7.4 | |
HEK293 | |
Immune Checkpoint Proteins |
25.9 kDa | |
Gly 24 - Lys 238 | |
LILRB4,ILT3,LIR5,CD85K,HM18 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |