Learn More
ACROBiosystems Mouse OX40 Ligand / TNFSF4 Protein, His Tag
Marke: ACROBiosystems TN4-M5241-50UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Tumor necrosis factor ligand superfamily member 4 (TNFSF4) is also known as glycoprotein Gp34, OX40 ligand (OX40L), TAX transcriptionally-activated glycoprotein 1 and CD252, which belongs to the tumor necrosis factor family. TNFSF4 is the ligand for CD134 and is expressed on such cells as DC2s (a subtype of dendritic cells) enabling amplification of Th2 cell differentiation. The interaction of TNFSF4-TNFSF4 is involved in the pathogenesis of multiple autoimmune and inflammatory diseases such as systemic lupus erythematosus (SLE), carotid artery disease and cancer. Furthermore, similar to other TNF superfamily members, membrane-bound OX40 Ligand (TNFSF4) exists as a homotrimer. Human TNFSF4 shares 46% amino acid sequence identity with its mouse counterpart.
This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 18.3 kDa. The protein migrates as 20 kDa and 22-25 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_033478.1 | |
50 μg | |
Gln 49 - Leu 198 | |
OX40L,TNFSF4,CD252,Glycoprotein Gp34,TXGP1,CD134 ligand,CD134L | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
18.3 kDa | |
Mouse O x 40 Ligand, His Tag (TN4-M5241) is expressed from human 293 cells (HEK293). It contains AA Gln 49 - Leu 198 (Accession # P43488-1). | |
-20°C | |
OX40 Ligand | |
Mouse | |
PBS, pH7.4 | |
HEK293 | |
Immune Checkpoint Proteins |