Learn More
ACROBiosystems Mouse OX40 / TNFRSF4 / CD134 Protein, His Tag
Marke: ACROBiosystems OX0-M5228-1MG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Tumor necrosis factor receptor superfamily member 4 (TNFRSF4) is also known as ACT35 antigen, OX40L receptor, TAX transcriptionally-activated glycoprotein 1 receptor, CD antigen CD134, OX40. OX40 / TNFRSF4 contains four TNFR-Cys repeats. TNFRSF4 is receptor for TNFSF4 / OX40L / GP34 and can interacts with TRAF2, TRAF3 and TRAF5.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 23.2 kDa. The protein migrates as 40-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_035789.1 | |
1 mg | |
Val 20 - Pro 211 | |
TNFRSF4,OX40,CD134,OX40L receptor,ACT35,TXGP1L | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
23.2 kDa | |
Mouse O x 40, His Tag (O x 0-M5228) is expressed from human 293 cells (HEK293). It contains AA Val 20 - Pro 211 (Accession # P47741-1). | |
-20°C | |
OX40 | |
Mouse | |
PBS, pH7.4 | |
HEK293 | |
Immune Checkpoint Proteins |