Learn More
ACROBiosystems Mouse PD-L1 / B7-H1 Protein, Mouse IgG2a Fc Tag, low endotoxin
low endotoxin
Marke:  ACROBiosystems PD1-M52A2-1MG
Weitere Details : Netto-Gewicht : 0.00001kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 15897487
  • 2634.48 € / 1 Milligramm
Verfügbar ab: 11-01-2023
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
Programmed cell death 1 ligand 1 (PDL1) is also known as B7-H, B7H1, MGC142294, MGC142296, PD-L1, PDCD1L1 and PDCD1LG1,which is a member of the growing B7 family of immune molecules and is involved in the regulation of cellular and humoral immune responses.PDL1 is a cell surface immunoglobulin superfamily with two Ig-like domains within the extracellular region and a short cytoplasmic domain. This protein is broadly expressed in the majority of peripheral tissues as well as hematopoietic cells. Interaction between PDL1 and its receptors belonging to the CD28 family of molecules provide both stimulatory and inhibitory signals in regulating T cell activation and tolerance. PDL1 may inhibit ongoing T-cell responses by inducing apoptosis and arresting cell-cycle progression.
This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 51.8 kDa. The protein migrates as 65-85 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spezifikation
Spezifikation
NP_068693
51.8 kDa
1 mg
Phe 19 - Thr 238
PD-L1,CD274,B7-H1,PDCD1L1,PDCD1LG1
Unconjugated
PBS, pH7.4
HEK293
Mouse
Powder
>95% as determined by SDS-PAGE
Mouse PD-L1, Mouse IgG2a Fc Tag, low endotoxin (PD1-M52A2) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Thr 238 (Accession # NP_068693).
-20°C
PD-L1
Mouse IgG Fc Tag
Less than 0.1 EU / µg by the LAL method.
Immune Checkpoint Proteins